missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorobenzonitrile 98.0+%, TCI America™

Catalog No. A19625G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19625G Supplier TCI America Supplier No. A19625G

Chemical Identifiers

CAS 21803-75-8
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00052917
InChI Key OREVCMGFYSUYPX-UHFFFAOYSA-N
Synonym 3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04
PubChem CID 519896
IUPAC Name 4-amino-3-chlorobenzonitrile
SMILES NC1=CC=C(C=C1Cl)C#N

Specifications

Melting Point 104°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 152.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Amino-3-chlorobenzonitrile