missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorobenzonitrile, 98%

Catalog No. AAA1215403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1215403 1 g
AAA1215406 5 g
AAA1215414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1215403 Supplier Thermo Scientific Chemicals Supplier No. A1215403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21803-75-8 | C7H5ClN2 | 152.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21803-75-8
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00052917
InChI Key OREVCMGFYSUYPX-UHFFFAOYSA-N
Synonym 3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04
PubChem CID 519896
IUPAC Name 4-amino-3-chlorobenzonitrile
SMILES NC1=CC=C(C=C1Cl)C#N

Specifications

Melting Point 98°C to 104°C
Quantity 1 g
UN Number UN3439
Beilstein 1817655
Formula Weight 152.58
Percent Purity 98%
Chemical Name or Material 4-Amino-3-chlorobenzonitrile
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only