missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. p-7054445
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6265403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6265403 Supplier Thermo Scientific Chemicals Supplier No. H6265403
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939968-08-8
Molecular Formula C12H16BF2NO2
Molecular Weight (g/mol) 255.072
MDL Number MFCD18434474
InChI Key JGOZEXIYNJERIP-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3,5-difluorophenyl boronic acid pinacol ester, 4-amino-3,5-difluorobenzeneboronic acid pinacol ester, 4-amino-3,5-difluorophenylboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxabor, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, benzenamine, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 59231931
IUPAC Name 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)F)N)F

Specifications

Color White
Quantity 1 g
Formula Weight 255.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Amino-3,5-difluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only