missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichloropyridine, 97%

Catalog No. p-7027734
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2472506 5 g
AAB2472514 25 g
AAB2472522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2472506 Supplier Thermo Scientific Chemicals Supplier No. B2472506

CAS: 22889-78-7 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22889-78-7
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00125023
InChI Key ISIQAMHROGZHOV-UHFFFAOYSA-N
Synonym 4-amino-3,5-dichloropyridine, 3,5-dichloro-4-aminopyridine, 3,5-dichloro-4-pyridinamine, 4-pyridinamine, 3,5-dichloro, 3,5-dichloro-4-amino pyridine, 3,5-dichloro-4-pyridine amine, 3,5-dichloro-pyridin-4-ylamine, 3,5-dichloro-4-pyridylamine, 3,5-dichloropyridin-4-ylamine, 4-amino-3,5-dichloro pyridine
PubChem CID 89888
IUPAC Name 3,5-dichloropyridin-4-amine
SMILES NC1=C(Cl)C=NC=C1Cl

Specifications

Melting Point 159°C to 163°C
Quantity 5 g
Beilstein 120547
Formula Weight 163.01
Percent Purity ≥98%
Chemical Name or Material 4-Amino-3,5-dichloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 245-304-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only