missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3,5-dichlorobenzonitrile 98.0+%, TCI America™
$29.83 - $74.56
Chemical Identifiers
| CAS | 78473-00-4 |
|---|---|
| Molecular Formula | C7H4Cl2N2 |
| Molecular Weight (g/mol) | 187.023 |
| MDL Number | MFCD00017341 |
| InChI Key | COFNCCWGWXFACE-UHFFFAOYSA-N |
| Synonym | 2,6-dichloro-4-cyanoaniline, 4-cyano-2,6-dichloroaniline, benzonitrile, 4-amino-3,5-dichloro, 3,5-dichloro-4-aminobenzonitrile, 4-amino-3,5-dichloro-benzonitrile, 4-amino-3,5-dichlorobenzenecarbonitrile, pubchem14453, acmc-1be7s, ksc381a4h, timtec-bb sbb003696 |
| PubChem CID | 522706 |
| IUPAC Name | 4-amino-3,5-dichlorobenzonitrile |
| SMILES | C1=C(C=C(C(=C1Cl)N)Cl)C#N |
Chemical Identifiers
| 78473-00-4 | |
| 187.023 | |
| COFNCCWGWXFACE-UHFFFAOYSA-N | |
| 522706 | |
| C1=C(C=C(C(=C1Cl)N)Cl)C#N |
| C7H4Cl2N2 | |
| MFCD00017341 | |
| 2,6-dichloro-4-cyanoaniline, 4-cyano-2,6-dichloroaniline, benzonitrile, 4-amino-3,5-dichloro, 3,5-dichloro-4-aminobenzonitrile, 4-amino-3,5-dichloro-benzonitrile, 4-amino-3,5-dichlorobenzenecarbonitrile, pubchem14453, acmc-1be7s, ksc381a4h, timtec-bb sbb003696 | |
| 4-amino-3,5-dichlorobenzonitrile |
Specifications
| 78473-00-4 | |
| White-Yellow | |
| MFCD00017341 | |
| 3439 | |
| COFNCCWGWXFACE-UHFFFAOYSA-N | |
| 4-amino-3,5-dichlorobenzonitrile | |
| 522706 | |
| ≥98.0% (GC) | |
| 4-Amino-3,5-dichlorobenzonitrile |
| 119°C | |
| C7H4Cl2N2 | |
| 1 g | |
| 2,6-dichloro-4-cyanoaniline, 4-cyano-2,6-dichloroaniline, benzonitrile, 4-amino-3,5-dichloro, 3,5-dichloro-4-aminobenzonitrile, 4-amino-3,5-dichloro-benzonitrile, 4-amino-3,5-dichlorobenzenecarbonitrile, pubchem14453, acmc-1be7s, ksc381a4h, timtec-bb sbb003696 | |
| C1=C(C=C(C(=C1Cl)N)Cl)C#N | |
| 187.023 | |
| 187.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No