missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3,5-dichlorobenzonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 119°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 187.02 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-3,5-dichlorobenzonitrile |
Safety and Handling
| TSCA | No |