missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichlorobenzonitrile 98.0+%, TCI America™

Catalog No. A19281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19281G 1 g
A19285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19281G Supplier TCI America Supplier No. A19281G
Only null left

Chemical Identifiers

CAS 78473-00-4
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.023
MDL Number MFCD00017341
InChI Key COFNCCWGWXFACE-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-cyanoaniline, 4-cyano-2,6-dichloroaniline, benzonitrile, 4-amino-3,5-dichloro, 3,5-dichloro-4-aminobenzonitrile, 4-amino-3,5-dichloro-benzonitrile, 4-amino-3,5-dichlorobenzenecarbonitrile, pubchem14453, acmc-1be7s, ksc381a4h, timtec-bb sbb003696
PubChem CID 522706
IUPAC Name 4-amino-3,5-dichlorobenzonitrile
SMILES C1=C(C=C(C(=C1Cl)N)Cl)C#N

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 187.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3,5-dichlorobenzonitrile