missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dibromopyridine 98.0+%, TCI America™

Catalog No. a23095g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23095G Supplier TCI America Supplier No. A23095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity reagent with 95% assay.

Chemical Identifiers

CAS 84539-34-4
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00234048
InChI Key MNRBVTDLMUNVLD-UHFFFAOYSA-N
PubChem CID 619792
IUPAC Name 3,5-dibromopyridin-4-amine
SMILES C1=C(C(=C(C=N1)Br)N)Br

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 251.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3,5-dibromopyridine