missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-picoline, 98+%, Thermo Scientific™

Catalog No. p-3593314
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393650010 1g Glass bottle
AC393650050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393650010 Supplier Thermo Scientific Chemicals Supplier No. 393650010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18437-58-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00186509
InChI Key GNCLPNMQEGMNTG-UHFFFAOYSA-N
Synonym 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118
PubChem CID 728670
IUPAC Name 2-methylpyridin-4-amine
SMILES CC1=NC=CC(=C1)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 1g
Formula Weight 108.14
Percent Purity 98+%
Chemical Name or Material 4-Amino-2-picoline
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

RUO – Research Use Only