missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-picoline, 98+%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 5g |
| Formula Weight | 108.14 |
| Percent Purity | 98+% |
| Chemical Name or Material | 4-Amino-2-picoline |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes serious eye damage. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately |
RUO – Research Use Only