missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methylpyridine 97.0+%, TCI America™

Catalog No. A23481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23481G Supplier TCI America Supplier No. A23481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versatile heterocyclic building block for chemical research applications.

Chemical Identifiers

CAS 18437-58-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00186509
InChI Key GNCLPNMQEGMNTG-UHFFFAOYSA-N
Synonym 4-amino-2-methylpyridine, 4-amino-2-picoline, 2-methyl-4-aminopyridine, 2-methyl-4-pyridinamine, 4-pyridinamine, 2-methyl, 2-methyl-pyridin-4-ylamine, 4 1h-pyridinimine, 2-methyl, 4-amino-picoline, 4-amino-6-picoline, pubchem1118
PubChem CID 728670
IUPAC Name 2-methylpyridin-4-amine
SMILES CC1=NC=CC(=C1)N

Specifications

Melting Point 96°C
Boiling Point 143°C
Quantity 1 g
Formula Weight 108.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methylpyridine