missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-methyl-1-butanol 98.0+%, TCI America™

Catalog No. p-7117527
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15775G 5 g
A157725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15775G Supplier TCI America Supplier No. A15775G
Only null left

Chemical Identifiers

CAS 44565-27-7
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD03093615
InChI Key DUAXLVGFFDFSAG-UHFFFAOYNA-N
PubChem CID 10129911
IUPAC Name 4-amino-2-methylbutan-1-ol
SMILES CC(CO)CCN

Specifications

Color Yellow
Quantity 5 g
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-2-methyl-1-butanol
EINECSNumber (2)-3927
TSCA No