missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-isopropyl-5-methylphenol hydrochloride, 97%

Catalog No. AAL1628203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1628203 1 g
AAL1628206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1628203 Supplier Thermo Scientific Chemicals Supplier No. L1628203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6321-11-5 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6321-11-5
MDL Number MFCD00035504

Specifications

Melting Point 258°C to 262°C
Odor Odorless
Molecular Formula C10H15ClNO
Quantity 1 g
Beilstein 3700651
Synonym 4-amino-2-isopropyl-5-methylphenol hydrochloride, 6-aminothymol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride, 4-amino-2-isopropyl-5-methyl-phenolhydrochloride, 4-amino-2-isopropyl-5-methyl-phenol hydrochloride, 4-amino-5-methyl-2-propan-2-yl phenol hydrochloride, phenol, 4-amino-5-methyl-2-1-methylethyl-, hydrochloride 1:1, 6-aminothymol hcl, 6-aminothymolhydrochlorid, 6-aminothymol hydrovhloride
InChI Key YLWORLAQZBRLSS-UHFFFAOYSA-N
SMILES CC1=C(C=C(C(=C1)O)C(C)C)N.Cl
IUPAC Name 4-amino-5-methyl-2-propan-2-ylphenol;hydrochloride
Molecular Weight (g/mol) 200.69
PubChem CID 80603
Formula Weight 201.7
Percent Purity 97%
Chemical Name or Material 4-Amino-2-isopropyl-5-methylphenol hydrochloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only