missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-fluorophenylboronic acid pinacol ester, 97%

Catalog No. AC444490010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444490010 1 g
AC444490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444490010 Supplier Thermo Scientific Chemicals Supplier No. 444490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 819057-45-9 | C12H17BFNO2 | 237.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 819057-45-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09951877
InChI Key FLMNWVXAEGUVNY-UHFFFAOYSA-N
Synonym 4-amino-2-fluorophenylboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-2-fluorobenzeneboronic acid pinacol ester, benzenamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-2-fluorophenyl boronic acid pinacol ester, 4-amino-2-fluorophenylboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl phenylamine, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-aniline
PubChem CID 44755207
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=C(N)C=C1

Specifications

Melting Point 163°C to 166°C
Color Brown to White
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 237.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-fluorophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only