missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine 98.0+%, TCI America™
$127.51 - $592.21
Chemical Identifiers
| CAS | 70700-44-6 |
|---|---|
| Molecular Formula | C6H9N5O2 |
| Molecular Weight (g/mol) | 183.171 |
| MDL Number | MFCD00059767 |
| InChI Key | WQJLLOCUCJRMMW-UHFFFAOYSA-N |
| Synonym | 6-Amino-2-dimethylamino-5-nitroso-4-pyrimidinol, 4-Hydroxy-2-dimethylamino-5-nitroso-6-aminopyrimidine |
| PubChem CID | 319506 |
| IUPAC Name | 6-amino-2-(dimethylamino)-5-nitroso-1H-pyrimidin-4-one |
| SMILES | CN(C)C1=NC(=O)C(=C(N1)N)N=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H0562100MG
|
TCI America
H0562100MG |
100 mg |
Each for $127.51
|
|
|||||
|
H05621G
|
TCI America
H05621G |
1 g |
Each for $592.21
|
|
|||||
Chemical Identifiers
| 70700-44-6 | |
| 183.171 | |
| WQJLLOCUCJRMMW-UHFFFAOYSA-N | |
| 319506 | |
| CN(C)C1=NC(=O)C(=C(N1)N)N=O |
| C6H9N5O2 | |
| MFCD00059767 | |
| 6-Amino-2-dimethylamino-5-nitroso-4-pyrimidinol, 4-Hydroxy-2-dimethylamino-5-nitroso-6-aminopyrimidine | |
| 6-amino-2-(dimethylamino)-5-nitroso-1H-pyrimidin-4-one |
Specifications
| 70700-44-6 | |
| C6H9N5O2 | |
| 100 mg | |
| WQJLLOCUCJRMMW-UHFFFAOYSA-N | |
| 6-amino-2-(dimethylamino)-5-nitroso-1H-pyrimidin-4-one | |
| 319506 | |
| ≥98.0% (T) | |
| 4-Amino-2-dimethylamino-6-hydroxy-5-nitrosopyrimidine [for Determination of Co(III), Fe(II)] |
| Purple-Red | |
| MFCD00059767 | |
| 6-Amino-2-dimethylamino-5-nitroso-4-pyrimidinol, 4-Hydroxy-2-dimethylamino-5-nitroso-6-aminopyrimidine | |
| CN(C)C1=NC(=O)C(=C(N1)N)N=O | |
| 183.171 | |
| 183.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No