missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloropyrimidine 98.0+%, TCI America™

Catalog No. p-7118039
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27191G 1 g
A27195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27191G Supplier TCI America Supplier No. A27191G
Only null left

Chemical Identifiers

CAS 7461-50-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00194051
InChI Key LPBDZVNGCNTELM-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyrimidine, 2-chloro-4-aminopyrimidine, 2-chloro-4-pyrimidinamine, 2-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 2-chloro, 2-chloropyrimidin-4-ylamine, 2-chloro-4-amino-pyrimidine, 2-chloropyrimidine-4-ylamine, zlchem 116, pubchem6955
PubChem CID 345752
IUPAC Name 2-chloropyrimidin-4-amine
SMILES NC1=CC=NC(Cl)=N1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloropyrimidine