missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloropyrimidine, 98%

Catalog No. p-7037207
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5587503 1 g
AAH5587506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5587503 Supplier Thermo Scientific Chemicals Supplier No. H5587503
Only null left

CAS: 7461-50-9 | C4H4ClN3 | 129.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7461-50-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00194051
InChI Key LPBDZVNGCNTELM-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyrimidine, 2-chloro-4-aminopyrimidine, 2-chloro-4-pyrimidinamine, 2-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 2-chloro, 2-chloropyrimidin-4-ylamine, 2-chloro-4-amino-pyrimidine, 2-chloropyrimidine-4-ylamine, zlchem 116, pubchem6955
PubChem CID 345752
IUPAC Name 2-chloropyrimidin-4-amine
SMILES NC1=CC=NC(Cl)=N1

Specifications

Melting Point 197°C to 205°C
Quantity 1 g
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 4-Amino-2-chloropyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only