missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chloropyridine, 98%

Catalog No. AAB2209806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2209806 5 g
AAB2209814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2209806 Supplier Thermo Scientific Chemicals Supplier No. B2209806
Only null left

CAS: 14432-12-3 | C5H5ClN2 | 128.559 g/mol

2-Chloropyridin-4-amine (14432-12-3) is used as a medicine, pigment intermediates. It is an additive of plant growth regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14432-12-3
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00060089
InChI Key BLBDTBCGPHPIJK-UHFFFAOYSA-N
Synonym 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine
PubChem CID 84432
IUPAC Name 2-chloropyridin-4-amine
SMILES C1=CN=C(C=C1N)Cl

Specifications

Melting Point 91°C to 94°C
Boiling Point 153°C (5 mmHg)
Packaging Plastic bottle
Quantity 5 g
Beilstein 108671
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-chloropyridine
Hazard Category H301+H311+H331-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 238-403-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only