missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-(chloromethyl)pyrimidine, 96%

Catalog No. aah6676003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6676003 1 g
AAH66760MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6676003 Supplier Thermo Scientific Chemicals Supplier No. H6676003

CAS: 79651-35-7 | C5H6ClN3 | 143.574 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79651-35-7
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD10697353
InChI Key JWAQENUKTAOJST-UHFFFAOYSA-N
Synonym 2-chloromethyl pyrimidin-4-amine, 2-chloromethyl-4-amino-pyrimidine, 4-pyrimidinamine, 2-chloromethyl, 2-chloromethyl-4-aminopyrimidine, 4-pyrimidinamine, 2-chloromethyl-9ci, 2-chloromethyl-pyrimidin-4-ylamine, 4-amino-2-chloromethylpyrimidine, 2-chloromethyl 4 amino pyrimidine, 2-chloromethyl-4-pyrimidinamine, 4-pyrimidinamine,2-chloromethyl-9ci
PubChem CID 21298827
IUPAC Name 2-(chloromethyl)pyrimidin-4-amine
SMILES C1=CN=C(N=C1N)CCl

Specifications

Color Colorless to Yellow
Density 1.35 g/mL
Boiling Point 264°C
Quantity 1 g
Formula Weight 143.57
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Amino-2-(chloromethyl)pyrimidine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only