missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7118071
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27845G 5 g
A278425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27845G Supplier TCI America Supplier No. A27845G
Only null left

Chemical Identifiers

CAS 20925-27-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00035926
InChI Key ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanoaniline, 2-chloro-4-aminobenzonitrile, benzonitrile, 4-amino-2-chloro, 4-amino-2-chloro-benzonitrile, 4-amino-2-chlorobenzenecarbonitrile, pubchem4620, 4-cyano-3-chloroaniline, acmc-1cfb7, 3-chloro-4-cyanobenzenamine, ksc497i5p
PubChem CID 88728
IUPAC Name 4-amino-2-chlorobenzonitrile
SMILES NC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 117°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 152.58
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Amino-2-chlorobenzonitrile