missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzonitrile, 97+%

Catalog No. AAL1592406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1592406 5 g
AAL1592414 25 g
AAL1592422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1592406 Supplier Thermo Scientific Chemicals Supplier No. L1592406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20925-27-3 | C7H5ClN2 | 152.58 g/mol

4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

Notes
Incompatible with acid chlorides, acid anhydrides and chloroformates.

Chemical Identifiers

CAS 20925-27-3
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00035926
InChI Key ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Synonym 3-chloro-4-cyanoaniline, 2-chloro-4-aminobenzonitrile, benzonitrile, 4-amino-2-chloro, 4-amino-2-chloro-benzonitrile, 4-amino-2-chlorobenzenecarbonitrile, pubchem4620, 4-cyano-3-chloroaniline, acmc-1cfb7, 3-chloro-4-cyanobenzenamine, ksc497i5p
PubChem CID 88728
IUPAC Name 4-amino-2-chlorobenzonitrile
SMILES NC1=CC=C(C#N)C(Cl)=C1

Specifications

Melting Point 116°C to 119°C
Quantity 5 g
UN Number UN3439
Beilstein 2936425
Formula Weight 152.58
Percent Purity ≥98%
Chemical Name or Material 4-Amino-2-chlorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 244-114-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only