missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2-chloro-3-nitropyridine 97.0+%, TCI America™
$121.16 - $365.35
Chemical Identifiers
| CAS | 2789-25-5 |
|---|---|
| Molecular Formula | C5H4ClN3O2 |
| Molecular Weight (g/mol) | 173.556 |
| MDL Number | MFCD07368898 |
| InChI Key | PDQAWJXOYURKPI-UHFFFAOYSA-N |
| PubChem CID | 10965014 |
| IUPAC Name | 2-chloro-3-nitropyridin-4-amine |
| SMILES | C1=CN=C(C(=C1N)[N+](=O)[O-])Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2798200MG
|
TCI America
A2798200MG |
200 mg |
Each for $121.16
|
|
|||||
|
A27981G
|
TCI America
A27981G |
1 g |
Each for $365.35
|
|
|||||
Chemical Identifiers
| 2789-25-5 | |
| 173.556 | |
| PDQAWJXOYURKPI-UHFFFAOYSA-N | |
| 2-chloro-3-nitropyridin-4-amine |
| C5H4ClN3O2 | |
| MFCD07368898 | |
| 10965014 | |
| C1=CN=C(C(=C1N)[N+](=O)[O-])Cl |
Specifications
| 2789-25-5 | |
| Yellow | |
| MFCD07368898 | |
| PDQAWJXOYURKPI-UHFFFAOYSA-N | |
| 2-chloro-3-nitropyridin-4-amine | |
| 10965014 | |
| ≥97.0% (GC,T) | |
| 4-Amino-2-chloro-3-nitropyridine |
| 187°C | |
| C5H4ClN3O2 | |
| 200 mg | |
| C1=CN=C(C(=C1N)[N+](=O)[O-])Cl | |
| 173.556 | |
| 173.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No