missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromopyrimidine-5-carbonitrile, 97%

Catalog No. p-7035071
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5277603 1 g
AAH5277606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5277603 Supplier Thermo Scientific Chemicals Supplier No. H5277603
Only null left

CAS: 94741-70-5 | C5H3BrN4 | 199.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94741-70-5
Molecular Formula C5H3BrN4
Molecular Weight (g/mol) 199.011
MDL Number MFCD00173661
InChI Key CXYLLFGNJJCGHM-UHFFFAOYSA-N
Synonym 4-amino-2-bromo-5-cyanopyrimidine, 4-amino-2-bromo-5-pyrimidinecarbonitrile, 5-pyrimidinecarbonitrile, 4-amino-2-bromo, 4-azanyl-2-bromanyl-pyrimidine-5-carbonitrile, 2-bromo-4-amino-5-cyanopyrimidine, acmc-209xix, 4-amino-2-bromo-pyrimidine-5-carbonitrile, 4-amino-2-bromopyrimidine-5-carbonitrile
PubChem CID 2735285
IUPAC Name 4-amino-2-bromopyrimidine-5-carbonitrile
SMILES C1=C(C(=NC(=N1)Br)N)C#N

Specifications

Melting Point 254°C (decomposition)
Quantity 1 g
Formula Weight 199.01
Percent Purity 97%
Chemical Name or Material 4-Amino-2-bromopyrimidine-5-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only