missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromopyridine, 97%

Catalog No. AAL2000706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2000706 5 g
AAL2000703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2000706 Supplier Thermo Scientific Chemicals Supplier No. L2000706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7598-35-8 | C5H5BrN2 | 173.01 g/mol

4-Amino-2-bromopyridine is used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-bromopyridine is used as medical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 7598-35-8
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD01646061
InChI Key GNTGEMWEXKBWBX-UHFFFAOYSA-N
Synonym 4-amino-2-bromopyridine, 2-bromo-4-aminopyridine, 2-bromo-4-pyridylamine, 2-bromo-4-pyridinamine, 4-pyridinamine, 2-bromo, 2-bromo-pyridin-4-ylamine, pubchem1100, pubchem1222, 2-brompyridin-4-amin, 2-bromopyridine-4-amine
PubChem CID 346455
IUPAC Name 2-bromopyridin-4-amine
SMILES NC1=CC=NC(Br)=C1

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 108928
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 4-Amino-2-bromopyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only