missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromopyridine, 95%

Catalog No. AC437410050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437410050 5 g Glass bottle
AC437410010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437410050 Supplier Thermo Scientific Chemicals Supplier No. 437410050

CAS: 7598-35-8 | C5H5BrN2 | 173.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7598-35-8
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.01
MDL Number MFCD01646061
InChI Key GNTGEMWEXKBWBX-UHFFFAOYSA-N
Synonym 4-amino-2-bromopyridine, 2-bromo-4-aminopyridine, 2-bromo-4-pyridylamine, 2-bromo-4-pyridinamine, 4-pyridinamine, 2-bromo, 2-bromo-pyridin-4-ylamine, pubchem1100, pubchem1222, 2-brompyridin-4-amin, 2-bromopyridine-4-amine
PubChem CID 346455
IUPAC Name 2-bromopyridin-4-amine
SMILES NC1=CC=NC(Br)=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 98.0°C to 100.0°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 173.01
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Amino-2-bromopyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only