missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyrimidine 98.0+%, TCI America™

Catalog No. A20915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20915G 5 g
A209125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20915G Supplier TCI America Supplier No. A20915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 461-98-3
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.159
MDL Number MFCD00006106
InChI Key BJJDXAFKCKSLTE-UHFFFAOYSA-N
Synonym 4-amino-2,6-dimethylpyrimidine, kyanmethin, cyanomethine, cyanmethine, kyanmethine, 2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-6-aminopyrimidine, 6-amino-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinylamine, 4-pyrimidinamine, 2,6-dimethyl
PubChem CID 68039
IUPAC Name 2,6-dimethylpyrimidin-4-amine
SMILES CC1=CC(=NC(=N1)C)N

Specifications

Melting Point 185°C
Color White-Yellow
Boiling Point 254°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2,6-dimethylpyrimidine