missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,6-dimethylpyrimidine, 99%

CAS: 461-98-3 | C6H9N3 | 123.159 g/mol

Supplier:  Thermo Scientific Chemicals B2217509

 View more versions of this product

Catalog No. AAB2217509

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Amino-2,6-dimethylpyrimidine
184°C to 186°C
MFCD00006106
14,5326
BJJDXAFKCKSLTE-UHFFFAOYSA-N
2,6-dimethylpyrimidin-4-amine
68039
99%
461-98-3
C6H9N3
10 g
4-amino-2,6-dimethylpyrimidine, kyanmethin, cyanomethine, cyanmethine, kyanmethine, 2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-6-aminopyrimidine, 6-amino-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinylamine, 4-pyrimidinamine, 2,6-dimethyl
CC1=CC(=NC(=N1)C)N
123.159
123.16

Chemical Identifiers

461-98-3
123.159
BJJDXAFKCKSLTE-UHFFFAOYSA-N
68039
CC1=CC(=NC(=N1)C)N
C6H9N3
MFCD00006106
4-amino-2,6-dimethylpyrimidine, kyanmethin, cyanomethine, cyanmethine, kyanmethine, 2,6-dimethyl-4-pyrimidinamine, 2,4-dimethyl-6-aminopyrimidine, 6-amino-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinylamine, 4-pyrimidinamine, 2,6-dimethyl
2,6-dimethylpyrimidin-4-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-320-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only