missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,6-dimethylpyridine 98.0+%, TCI America™
$209.84 - $833.61
Chemical Identifiers
| CAS | 3512-80-9 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00130078 |
| InChI Key | ZJXMKPARTVOUAM-UHFFFAOYSA-N |
| PubChem CID | 77047 |
| IUPAC Name | 2,6-dimethylpyridin-4-amine |
| SMILES | CC1=CC(=CC(=N1)C)N |
Chemical Identifiers
| 3512-80-9 | |
| 122.171 | |
| ZJXMKPARTVOUAM-UHFFFAOYSA-N | |
| 2,6-dimethylpyridin-4-amine |
| C7H10N2 | |
| MFCD00130078 | |
| 77047 | |
| CC1=CC(=CC(=N1)C)N |
Specifications
| 3512-80-9 | |
| White-Yellow | |
| C7H10N2 | |
| 1 g | |
| ZJXMKPARTVOUAM-UHFFFAOYSA-N | |
| 2,6-dimethylpyridin-4-amine | |
| 77047 | |
| ≥98.0% (GC) | |
| 4-Amino-2,6-dimethylpyridine |
| 193°C | |
| 246°C | |
| MFCD00130078 | |
| 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine | |
| CC1=CC(=CC(=N1)C)N | |
| 122.171 | |
| 122.17 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0670
TSCA : No