missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,6-dimethylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 193°C |
| Color | White-Yellow |
| Boiling Point | 246°C |
| Quantity | 1 g |
| Synonym | 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine |
| Formula Weight | 122.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Amino-2,6-dimethylpyridine |
Safety and Handling
| EINECSNumber | (9)-0670 |
| TSCA | No |