Learn More
4-Amino-2,6-dimethylpyridine, 98%
CAS: 3512-80-9 | C7H10N2 | 122.171 g/mol
$175.81 - $723.78
Chemical Identifiers
| CAS | 3512-80-9 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00130078 |
| InChI Key | ZJXMKPARTVOUAM-UHFFFAOYSA-N |
| PubChem CID | 77047 |
| IUPAC Name | 2,6-dimethylpyridin-4-amine |
| SMILES | CC1=CC(=CC(=N1)C)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6197003
|
Thermo Scientific Chemicals
H6197003 |
1 g |
Each for $175.81
|
|
|||||
|
AAH6197006
|
Thermo Scientific Chemicals
H6197006 |
5 g |
Each for $723.78
|
|
|||||
Description
4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-2,6-dimethylpyridine, is an important raw material and intermediate used in pharmaceuticals, agrochemicals, dyestuff, field.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 3512-80-9 | |
| 122.171 | |
| ZJXMKPARTVOUAM-UHFFFAOYSA-N | |
| 2,6-dimethylpyridin-4-amine |
| C7H10N2 | |
| MFCD00130078 | |
| 77047 | |
| CC1=CC(=CC(=N1)C)N |
Specifications
| 3512-80-9 | |
| MFCD00130078 | |
| UN2811 | |
| Slightly soluble in water. | |
| CC1=CC(=CC(=N1)C)N | |
| 122.171 | |
| 122.17 | |
| Solid |
| C7H10N2 | |
| 1 g | |
| 4-amino-2,6-dimethylpyridine, 2,6-dimethyl-pyridin-4-ylamine, 2,6-dimethylpyridin-4-ylamine, 4-amino-2,6-lutidine, 4-pyridinamine, 2,6-dimethyl, 2,6-dimethylpyridin-4yl-amine, 2,6-dimethyl-4-pyridinamine, 2,6-dimethyl-4-aminopyridine, pubchem19474, 4-amino-2, 6-lutidine | |
| ZJXMKPARTVOUAM-UHFFFAOYSA-N | |
| 2,6-dimethylpyridin-4-amine | |
| 77047 | |
| 98% | |
| 4-Amino-2,6-dimethylpyridine |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 225-515-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only