missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical

CAS: 14691-88-4 | C9H19N2O | 171.26 g/mol

$250.88 - $805.16

Chemical Identifiers

CAS 14691-88-4
Molecular Formula C9H19N2O
Molecular Weight (g/mol) 171.26
MDL Number MFCD00006479
Synonym 4-Amino-TEMPO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172730010
SDS View Documents
Thermo Scientific Chemicals
172730010
1 g Glass bottle
Each for $250.88
Only null left
 
AC172730050
SDS View Documents
Thermo Scientific Chemicals
172730050
5 g Glass bottle
Each for $805.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Polymerization reagent
Specifications

Chemical Identifiers

14691-88-4
171.26
4-Amino-TEMPO
C9H19N2O
MFCD00006479

Specifications

14691-88-4
100.0
96% min. (GC)
C9H19N2O
1 g
Solubility in water: soluble. Other solubilities: soluble in ethanol,chloroform,hexane
171.26
Adhering Crystals, Needles or Moist Mass
96.0
Authentic
Glass bottle
MFCD00006479
4-Amino-TEMPO
171.26
97%
4-Amino-2,2,6,6-tetramethylpiperidinooxy
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 238-738-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only