missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,2,6,6-tetramethylpiperidine 98.0+%, TCI America™

Catalog No. p-7117230
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A107725ML 25 mL
A1077500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A107725ML Supplier TCI America Supplier No. A107725ML
Only null left

Chemical Identifiers

CAS 36768-62-4
Molecular Formula C9H22N2
Molecular Weight (g/mol) 158.29
MDL Number MFCD00005984
InChI Key FTVFPPFZRRKJIH-UHFFFAOYSA-P
Synonym 4-amino-2,2,6,6-tetramethylpiperidine, triacetonediamine, 2,2,6,6-tetramethyl-4-aminopiperidine, 4-piperidinamine, 2,2,6,6-tetramethyl, unii-etf220q65r, 2,2,6,6-tetramethyl-4-piperidylamine, 4-amino-2,2,6,6-tetramethyl-piperidine, piperidine, 4-amino-2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl-4-piperidinamine, 4-amino-2,2,6,6-tetramethyl piperidine
PubChem CID 37524
IUPAC Name 4-azaniumyl-2,2,6,6-tetramethylpiperidin-1-ium
SMILES CC1(C)CC([NH3+])CC(C)(C)[NH2+]1

Specifications

Color Yellow
pH 12.3 (99g/L H2O soln. 20°C)
Boiling Point 93°C
Quantity 25 mL
UN Number 2735
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidine
EINECSNumber (5)-6089
RTECSNumber TM4290100
TSCA Yes