missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2,2,6,6-tetramethylpiperidine, 98%

Catalog No. AAB2406206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2406206 5 g
AAB2406214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2406206 Supplier Thermo Scientific Chemicals Supplier No. B2406206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36768-62-4 | C9H22N2 | 158.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36768-62-4
Molecular Formula C9H22N2
Molecular Weight (g/mol) 158.29
MDL Number MFCD00005984
InChI Key FTVFPPFZRRKJIH-UHFFFAOYSA-P
Synonym 4-amino-2,2,6,6-tetramethylpiperidine, triacetonediamine, 2,2,6,6-tetramethyl-4-aminopiperidine, 4-piperidinamine, 2,2,6,6-tetramethyl, unii-etf220q65r, 2,2,6,6-tetramethyl-4-piperidylamine, 4-amino-2,2,6,6-tetramethyl-piperidine, piperidine, 4-amino-2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl-4-piperidinamine, 4-amino-2,2,6,6-tetramethyl piperidine
PubChem CID 37524
SMILES CC1(C)CC([NH3+])CC(C)(C)[NH2+]1

Specifications

Melting Point 16°C to 18°C
Density 0.912
Boiling Point 188°C to 189°C
Flash Point 75°C (167°F)
Quantity 5 g
IUPAC Name 4-azaniumyl-2,2,6,6-tetramethylpiperidin-1-ium
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidine
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 253-197-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only