missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 97.0+%, TCI America™
$171.49 - $516.35
Chemical Identifiers
| CAS | 14691-88-4 |
|---|---|
| Molecular Formula | C9H19N2O |
| MDL Number | MFCD00006479 |
| Synonym | 4-Amino-TEMPO |
Chemical Identifiers
| 14691-88-4 | |
| C9H19N2O | |
| MFCD00006479 | |
| 4-Amino-TEMPO |
Specifications
| 14691-88-4 | |
| C9H19N2O | |
| 1 g | |
| 171.26 | |
| Crystalline Lumps |
| 98°C | |
| MFCD00006479 | |
| 4-Amino-TEMPO | |
| ≥97.0% (GC) | |
| 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator