missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 98°C |
| Quantity | 5 g |
| Formula Weight | 171.26 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |