missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-tert-butoxycarbonylpiperidine 97.0+%, TCI America™

Catalog No. p-7117695
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A20451G 1 g
A20455G 5 g
2 options

Catalog No. A20451G

Supplier: TCI America A20451G

Only null left

Chemical Identifiers

CAS 87120-72-7
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD01076201
InChI Key LZRDHSFPLUWYAX-UHFFFAOYSA-O
Synonym 4-amino-1-boc-piperidine, 1-boc-4-aminopiperidine, n-boc-4-aminopiperidine, n-boc-4-amino-piperidine, 4-amino-1-n-boc-piperidine, 4-amino-n-boc piperidine, 1-boc-4-amino piperidine, 1-n-boc-4-aminopiperidine, 1-t-butoxycarbonyl-4-aminopiperidine, n-1-boc-4-aminopiperidine
PubChem CID 1268291
IUPAC Name 1-[(tert-butoxy)carbonyl]piperidin-4-aminium
SMILES CC(C)(C)OC(=O)N1CCC([NH3+])CC1

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1-tert-butoxycarbonylpiperidine