missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-methyl-1H-pyrazole, 95%

Catalog No. AAH3372806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3372806 5 g
AAH3372803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3372806 Supplier Thermo Scientific Chemicals Supplier No. H3372806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69843-13-6 | C4H7N3 | 97.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69843-13-6
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD05667135
InChI Key LBGSWBJURUFGLR-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-4-amine, 1-methyl-1h-pyrazol-4-ylamine, 4-amino-1-methylpyrazole, 1h-pyrazol-4-amine, 1-methyl, 1-methyl-1h-pyrazole-4-ylamine, 4-amino-1-methyl-1h-pyrazole, 1-methylpyrazole-4-ylamine, acmc-20a0x1, 4-amino-1-methyl-pyrazole, 1-methyl-1h-pyrazole-4-amine
PubChem CID 4770990
IUPAC Name 1-methylpyrazol-4-amine
SMILES CN1C=C(C=N1)N

Specifications

Melting Point 47°C to 50°C
Quantity 5 g
Beilstein 4368862
Sensitivity Air Sensitive
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 4-Amino-1-methyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only