missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-diethylaminopentane 98.0+%, TCI America™

Catalog No. D100425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D100425ML Supplier TCI America Supplier No. D100425ML
Only null left

Specifications

CAS 140-80-7
Color Yellow
Boiling Point 101°C
Molecular Formula C9H22N2
Quantity 25 mL
MDL Number MFCD00008091
UN Number 2946
Synonym N1,N1-Diethyl-1,4-diaminopentane, N1,N1-Diethyl-1,4-pentanediamine
InChI Key CAPCBAYULRXQAN-UHFFFAOYSA-N
SMILES CCN(CC)CCCC(C)N
IUPAC Name 1-N,1-N-diethylpentane-1,4-diamine
Molecular Weight (g/mol) 158.289
PubChem CID 78953
Formula Weight 158.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-diethylaminopentane
Show More Show Less
RTECSNumber XE1900000
TSCA Yes