missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-butanol 98.0+%, TCI America™

Catalog No. p-7117187
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A10135G 5 g
A101325G 25 g
2 options

Catalog No. A10135G

Supplier: TCI America A10135G

Only null left

Chemical Identifiers

CAS 13325-10-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008230
InChI Key BLFRQYKZFKYQLO-UHFFFAOYSA-N
Synonym 4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
PubChem CID 25868
IUPAC Name 4-aminobutan-1-ol
SMILES C(CCO)CN

Specifications

Color Yellow
Boiling Point 206°C
Quantity 5 g
UN Number 2735
Formula Weight 89.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-butanol