missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-Boc-piperidine-4-carboxylic acid, 98%

Catalog No. p-7047812
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1979403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1979403 Supplier Thermo Scientific Chemicals Supplier No. L1979403

CAS: 183673-71-4 | C11H20N2O4 | 244.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 183673-71-4
Molecular Formula C11H20N2O4
Molecular Weight (g/mol) 244.29
MDL Number MFCD01318728
InChI Key YNHLVALLAURVJF-UHFFFAOYSA-N
Synonym 1-boc-4-aminopiperidine-4-carboxylic acid, 4-amino-1-boc-piperidine-4-carboxylic acid, 4-amino-1-boc-isonipecotic acid, 4-amino-1-tert-butoxycarbonyl piperidine-4-carboxylic acid, h-pip boc-oh, 4,4-acp 1-n-boc, 4-amino-piperidine-1,4-dicarboxylic acid mono-tert-butyl ester, 1-boc-4-amino-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxy carbonyl piperidine-4-carboxylic acid
PubChem CID 693797
SMILES CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O

Specifications

Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 7588844
IUPAC Name 4-amino-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-4-carboxylic acid
Formula Weight 244.29
Percent Purity 98%
Chemical Name or Material 4-Amino-1-Boc-piperidine-4-carboxylic acid
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only