missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-benzylpiperidine 98.0+%, TCI America™

Catalog No. A108625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A108625G 25 g
A1086100G 100 g
A1086500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A108625G Supplier TCI America Supplier No. A108625G

Chemical Identifiers

CAS 50541-93-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00006504
InChI Key YUBDLZGUSSWQSS-UHFFFAOYSA-N
Synonym 4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
PubChem CID 415852
IUPAC Name 1-benzylpiperidin-4-amine
SMILES C1CN(CCC1N)CC2=CC=CC=C2

Specifications

Color Yellow
Quantity 25 g
Formula Weight 190.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amino-1-benzylpiperidine