missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-benzylpiperidine, 98%

Catalog No. p-55653
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187660050 5 g Glass bottle
AC187660250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC187660050 Supplier Thermo Scientific Chemicals Supplier No. 187660050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50541-93-0 | C12H18N2 | 190.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50541-93-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00006504
InChI Key YUBDLZGUSSWQSS-UHFFFAOYSA-N
Synonym 4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
PubChem CID 415852
IUPAC Name 1-benzylpiperidin-4-amine
SMILES C1CN(CCC1N)CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 152°C (9.7 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5410 to 1.543
Quantity 5 g
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether
Formula Weight 190.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Amino-1-benzylpiperidine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 256-620-9

RUO – Research Use Only