missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,8-naphthalimide, 95%

Catalog No. p-7042918
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64358MC 100 mg
AAJ6435803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64358MC Supplier Thermo Scientific Chemicals Supplier No. J64358MC
Only null left

CAS: 1742-95-6 | C12H8N2O2 | 212.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1742-95-6
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.21
MDL Number MFCD00006921
InChI Key SSMIFVHARFVINF-UHFFFAOYSA-N
Synonym 4-amino-1,8-naphthalimide, 4-aminonaphthalimide, 6-amino-benzo de isoquinoline-1,3-dione, 6-amino-1h-benzo de isoquinoline-1,3 2h-dione, 4-aminonaphthalene-1,8-dicarboximide, naphthalimide, 4-amino, dfp 1, 1h-benz de isoquinoline-1,3 2h-dione, 6-amino, 6-aminobenzo de isoquinoline-1,3-dione, parp inhibitor v, 4-ani
PubChem CID 1720
ChEBI CHEBI:40071
SMILES NC1=C2C=CC=C3C(=O)NC(=O)C(C=C1)=C23

Specifications

Melting Point >360°C
Color Orange
Density 1.105
Quantity 100 mg
Solubility Information Soluble in DMSO
IUPAC Name 6-aminobenzo[de]isoquinoline-1,3-dione
Formula Weight 212.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Amino-1,8-naphthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-110-1
RTECSNumber DE4081000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only