missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,3,5-trimethyl-1H-pyrazole, 97%

Catalog No. AAL1062506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1062506 5 g
AAL1062503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1062506 Supplier Thermo Scientific Chemicals Supplier No. L1062506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28466-21-9 | C6H11N3 | 125.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28466-21-9
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.175
MDL Number MFCD00052883
InChI Key SSDGMKHZMNTWLS-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazol-4-amine, 4-amino-1,3,5-trimethylpyrazole, trimethyl-1h-pyrazol-4-amine, pyrazole, 4-amino-1,3,5-trimethyl, 4-amino-1,3,5-trimethyl-1h-pyrazole, trimethylpyrazol-4-amine, 1,3,5-trimethyl-1h-pyrazol-4-ylamine, 1h-pyrazol-4-amine, 1,3,5-trimethyl, 1,3,5-trimethyl-1h-pyrazole-4-amine, 1,3,5-trimethylpyrazole-4-ylamine
PubChem CID 161603
IUPAC Name 1,3,5-trimethylpyrazol-4-amine
SMILES CC1=C(C(=NN1C)C)N

Specifications

Melting Point 99°C to 105°C
Quantity 5 g
Beilstein 110739
Formula Weight 125.17
Percent Purity 97%
Chemical Name or Material 4-Amino-1,3,5-trimethyl-1H-pyrazole
EINECSNumber 000-000-0
RTECSNumber UQ6125100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only