missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,4-triazole 98.0+%, TCI America™

Supplier:  TCI America A1137250G

 View more versions of this product

Catalog No. A1137250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Amino-1,2,4-triazole
86°C
MFCD00003099
4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
C1=NN=CN1N
84.082
84.08
Crystalline Powder
584-13-4
C2H4N4
250 g
FMCUPJKTGNBGEC-UHFFFAOYSA-N
1,2,4-triazol-4-amine
11432
≥98.0% (HPLC,T)

Chemical Identifiers

584-13-4
84.082
FMCUPJKTGNBGEC-UHFFFAOYSA-N
11432
C1=NN=CN1N
C2H4N4
MFCD00003099
4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
1,2,4-triazol-4-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0603

RTECSNumber : XZ3850200

TSCA : No

SDS
Documents

Documents

Promotions

Promotions