missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,4-triazole 98.0+%, TCI America™

Catalog No. A113725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A113725G 25 g
A1137250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A113725G Supplier TCI America Supplier No. A113725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00003099
InChI Key FMCUPJKTGNBGEC-UHFFFAOYSA-N
Synonym 4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
PubChem CID 11432
IUPAC Name 1,2,4-triazol-4-amine
SMILES C1=NN=CN1N

Specifications

Melting Point 86°C
Quantity 25 g
Formula Weight 84.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-1,2,4-triazole
EINECSNumber (5)-0603
RTECSNumber XZ3850200
TSCA No