missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1,2,4-triazole, 99%

Catalog No. AAA1771606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1771606 5 g
AAA1771614 25 g
AAA1771622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1771606 Supplier Thermo Scientific Chemicals Supplier No. A1771606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 584-13-4 | C2H4N4 | 84.082 g/mol

4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00003099
InChI Key FMCUPJKTGNBGEC-UHFFFAOYSA-N
Synonym 4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
PubChem CID 11432
IUPAC Name 1,2,4-triazol-4-amine
SMILES C1=NN=CN1N

Specifications

Melting Point 83°C to 88°C
Quantity 5 g
Beilstein 107563
Solubility Information Soluble in water and methanol.
Formula Weight 84.08
Percent Purity 99%
Chemical Name or Material 4-Amino-1,2,4-triazole
Hazard Category H315-H319-H350
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 209-533-5
RTECSNumber XZ3850200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only