missing translation for 'onlineSavingsMsg'
Learn More

4-Amidinopyridine hydrochloride, 98+%

Catalog No. AAA1105014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1105014 25 g
AAA1105006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1105014 Supplier Thermo Scientific Chemicals Supplier No. A1105014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6345-27-3 | C6H8ClN3 | 157.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6345-27-3
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.60
MDL Number MFCD00040385
InChI Key IONKMFGAXKCLMI-UHFFFAOYSA-N
Synonym isonicotinimidamide hydrochloride, pyridine-4-carboximidamide hydrochloride, 4-amidinopyridine hydrochloride, isonicotinamidine hydrochloride, pyridine-4-carboxamidine hydrochloride, 4-pyridinecarboximidamide hydrochloride, 4-pyridinecarboximidamide, monohydrochloride, isonicotinamidine, hydrochloride, 4-amidino-pyridine hydrochloride, pyridine-4-carboxamidine, chloride
PubChem CID 2776844
IUPAC Name pyridine-4-carboximidamide;hydrochloride
SMILES Cl.NC(=N)C1=CC=NC=C1

Specifications

Melting Point ∼245°C (decomposition)
Quantity 25 g
Beilstein 3562678
Sensitivity Hygroscopic
Formula Weight 157.6
Percent Purity ≥98%
Chemical Name or Material 4-Amidinopyridine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only