missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzaldehyde, 97%

CAS: 40663-68-1 | C10H10O2 | 162.19 g/mol

$100.79 - $300.10

Chemical Identifiers

CAS 40663-68-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00014133
InChI Key TYNJQOJWNMZQFZ-UHFFFAOYSA-N
Synonym 4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
PubChem CID 95942
IUPAC Name 4-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=C(C=O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1377309
View Documents
Thermo Scientific Chemicals
A1377309
10 g
Each for $100.79
Only null left
 
AAA1377318
View Documents
Thermo Scientific Chemicals
A1377318
50 g
Each for $300.10
Only null left
 
Description

Description

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.
Specifications

Chemical Identifiers

40663-68-1
162.19
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
95942
C=CCOC1=CC=C(C=O)C=C1
C10H10O2
MFCD00014133
4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
4-prop-2-enoxybenzaldehyde

Specifications

40663-68-1
150°C to 152°C (18 mmHg)
C10H10O2
MFCD00014133
2206924
4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
4-prop-2-enoxybenzaldehyde
95942
97%
1.058
>110°C (230°F)
1.5695
10 g
Air Sensitive
Immiscible in water.
C=CCOC1=CC=C(C=O)C=C1
162.19
162.19
4-Allyloxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c

H315-H317-H319

EINECSNumber : 255-027-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only