missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzaldehyde, 97%

Catalog No. AAA1377318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1377318 50 g
AAA1377309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1377318 Supplier Thermo Scientific Chemicals Supplier No. A1377318

CAS: 40663-68-1 | C10H10O2 | 162.19 g/mol

4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Allyloxybenzaldehyde is used as pharmaceutical intermediates.

Solubility
Immiscible in water.

Notes
Use with adequate ventilation. Store under inert atmosphere. Protect from moisture and heat.

Chemical Identifiers

CAS 40663-68-1
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00014133
InChI Key TYNJQOJWNMZQFZ-UHFFFAOYSA-N
Synonym 4-allyloxybenzaldehyde, 4-allyloxy benzaldehyde, p-allyloxy benzaldehyde, p-allyloxybenzaldehyde, benzaldehyde, 4-2-propenyloxy, 4-prop-2-en-1-yloxy benzaldehyde, 4-prop-2-enyloxybenzaldehyde, 4-allyloxybenzaldehyd, acmc-20ampt
PubChem CID 95942
IUPAC Name 4-prop-2-enoxybenzaldehyde
SMILES C=CCOC1=CC=C(C=O)C=C1

Specifications

Density 1.058
Boiling Point 150°C to 152°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5695
Quantity 50 g
Beilstein 2206924
Sensitivity Air Sensitive
Solubility Information Immiscible in water.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Allyloxybenzaldehyde
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
EINECSNumber 255-027-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only